Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(propylene glycol) propyl ether, mixture of isomers

Catalog No. 4842101L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-421-01L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-421-01L Supplier Sigma Aldrich Supplier No. 4842101L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product of Dow Chemical

Specifications

CAS 29911-27-1
Density 0.92 g/mL (at 25°C)
Boiling Point 212°C
Molecular Formula C9H20O3
Linear Formula CH3CH2CH2OC3H6OC3H6OH
Refractive Index n20/D 1.424 (literature)
Quantity 1 L
MDL Number MFCD00274589
Synonym DOWANOL™ DPnP
Molecular Weight (g/mol) 176.25
Percent Purity ≥98.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.