Learn More
Bioworld Di-Sodium Hydrogen Phosphate Dihydrate, 250 g
Supplier:  Bioworld 404000721
Encompass_Preferred
A reagent with high buffering capacitySpecifications:Synonym: Disodium Hydrogen Phosphate; Sorensen's PhosphateLoss on drying: 18.0 - 22.0%Arsenic (As): 3µg/gFluoride: 0.005%Insolubles: 0.2%Appearance: Granules
Catalog No. 50-255-851