Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

di-Sodium hydrogen phosphate anhydrous EMPROVE™ ESSENTIAL Ph Eur, BP, USP, JPE, E 339, 101.0%, MilliporeSigma™

Catalog No. M1065859013 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1065859013 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065859013 Supplier MilliporeSigma Supplier No. 1.06585.9013
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96

Specifications

Melting Point 250°C (decomposition)
pH 8.7 to 9.3 (10 g/L, H2O, 20°C)
Quantity 12 kg
Assay Percent Range Assay (calculated on dried substance according to Ph Eur): 98.0 to 101.0%; Assay (calculated on dried substance according to USP): 98.0 to 100.5%; Assay (acidimetric; dried substance): 99.0 to 101.0%
Solubility Information 77 g/L
Packaging Plastic Bucket
Density 880 kg/m3
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.