Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

di-Sodium Hydrogen Phosphate, Anhydrous, For Analysis, EMSURE™, ACS, MilliporeSigma™

Catalog No. M1065869012 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Plastic Bottle
Plastic Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065869012 12 kg Plastic Drum
M1065862500 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1065869012 Supplier MilliporeSigma Supplier No. 1065869012
Add to Cart
Add to Cart

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 250°C (decomposition)
Color White
pH 8.7 to 9.3 (10g/L, H2O, 20°C)
Physical Form Solid
Quantity 12 kg
Solubility Information 77g/L
Formula Weight 141.96
Percent Purity >99.0%
Packaging Plastic Drum
EINECSNumber 231-448-7
RTECSNumber WC4500000
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.