Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

di-Sodium Hydrogen Phosphate Heptahydrate, For Analysis, EMSURE™, ACS, MilliporeSigma™

Catalog No. M1065759025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065759025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065759025 Supplier MilliporeSigma Supplier No. 1.06575.9025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 48°C (Elimination of water of crystallization)
Color White
pH 8.7 to 9.3
Physical Form Solid
Quantity 25 kg
Solubility Information 154g/L
Formula Weight 268.03
Percent Purity 98.0 to 102.0%
Packaging Fiber Carton
Density 1.68 g/3 (20 °C)
EINECSNumber 231-448-7
RTECSNumber WC4600000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.