Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-T-Butyl Oxaziridine

Catalog No. 900624500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-062-4500MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 90-062-4500MG Supplier Sigma Aldrich Supplier No. 900624500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This oxaziridine has been used in synthetic methods for amination, heteroatom transfer, and C-H functionalization. Recently, the lab of Laszlo Kurti demonstrated its application for direct electrophilic primary and secondary amination of arylmetals in the presence of Cu(I) salts -- void of precious metal catalysts, ligands, protecting groups, or directing groups.Non-Deprotonative Primary and Secondary Amination of (Hetero)ArylmetalsRapid heteroatom transfer to arylmetals utilizing multifunctional reagent scaffoldsDiverse sp3 C-H functionalization through alcohol beta-sulfonyloxylation

Specifications

CAS 5905-52-2
Molecular Formula C6H10FeO6 -+ xH2O
Linear Formula [CH3CH(OH)COO]2Fe -+ xH2O
Quantity 100 g
Synonym Kurti oxaziridine
Molecular Weight (g/mol) 233.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.