Learn More
Strem, An Ascensus Company CAS# 13716-10-4. 5g. Di-t-butylchlorophosphine, min. 98%. MFCD00008815
SDP
Catalog No. 5090110003
Encompass
CAS# 13716-10-4. 5g. Di-t-butylchlorophosphine, min. 98%. MFCD00008815. Molecular Weight: 180.66. Molecular Formula: [(CH3)3C]2PCl. Color/form: colorless to light yellow liq. Strem# 15-1000. http://www.strem.com/catalog/v/15-1000/
Catalog No. 50-901-10003 Supplier Strem, An Ascensus Company Supplier No. 1510005G
CAS# 13716-10-4. 5g. Di-t-butylchlorophosphine, min. 98%. MFCD00008815. Molecular Weight: 180.66. Molecular Formula: [(CH3)3C]2PCl. Color/form: colorless to light yellow liq. Strem# 15-1000. http://www.strem.com/catalog/v/15-1000/