Learn More
Strem, An Ascensus Company CAS# 19429-30-2. 5g. Di-t-butyltin dichloride, 98%. MFCD00009854
SDP
Supplier:  Strem, An Ascensus Company 5011405G
Encompass
CAS# 19429-30-2. 5g. Di-t-butyltin dichloride, 98%. MFCD00009854. Molecular Weight: 303.83. Molecular Formula: (t-C4H9)2SnCl2. Color/form: white xtl. Strem# 50-1140. http://www.strem.com/catalog/v/50-1140/
Catalog No. 50-901-12590