Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl 3,3'-Iminodipropionate 97.0+%, TCI America™
SDP

Catalog No. D41101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41101G 1 g
D41105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41101G Supplier TCI America Supplier No. D41101G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 128988-04-5 | C14H27NO4 | 273.373 g/mol

Chemical Identifiers

CAS 128988-04-5
Molecular Formula C14H27NO4
Molecular Weight (g/mol) 273.373
MDL Number MFCD07781379
InChI Key KCTOWZYKZDFZMQ-UHFFFAOYSA-N
PubChem CID 10945622
IUPAC Name tert-butyl 3-[[3-[(2-methylpropan-2-yl)oxy]-3-oxopropyl]amino]propanoate
SMILES CC(C)(C)OC(=O)CCNCCC(=O)OC(C)(C)C

Specifications

Color Colorless
Synonym 3,3′C-Iminodipropionic Acid Di-tert-butyl Ester
Quantity 1 g
Formula Weight 273.37
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Di-tert-butyl 3,3′-Iminodipropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.