Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-Tert-Butyl Dicarbonate

Catalog No. 34660100GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-660-100GF 100 g
34-660-500GF 500 g
34-660-25GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 34-660-100GF Supplier Sigma Aldrich Supplier No. 34660100GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di-Tert-Butyl Dicarbonate

Specifications

CAS 24424-99-5
Melting Point 23°C
Density 0.95 g/mL at 25°C
Boiling Point 56°C to 57°C (0.5 mmHg, lit.)
Molecular Formula C10H18O5
Refractive Index n20/D 1.409
Linear Formula [(CH3)3COCO]2O
MDL Number MFCD00008805
Quantity 100 g
Synonym Boc2O; Boc anhydride; Di-tert-butyl pyrocarbonate
Molecular Weight (g/mol) 218.25 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.