Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl Dicarbonate 95.0+%, TCI America™
SDP

Catalog No. D1547100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1547100G 100 g
D154725G 25 g
D1547500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1547100G Supplier TCI America Supplier No. D1547100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.249
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Color White-Yellow
Quantity 100 g
UN Number 2930
Formula Weight 218.25
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Di-tert-butyl Dicarbonate [Boc-reagent for Amino Acid]
EINECSNumber (2)-3415
RTECSNumber HT0230000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.