Learn More
Medchemexpress LLC Di-tert-butyl diisopropylphosphoramidite | 137348-86-8 | 153506 | 96.9% | 277.39 | 10 MM / 1 ML
SDP
Catalog No. 5000451324
Encompass_Preferred
Di-tert-butyl diisopropylphosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides.
  • It appears as a liquid, colorless to light yellow.
  • It ships at room temperature in the continental US; conditions may vary elsewhere.
  • Store at 4°C under nitrogen.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen.
Catalog No. 50-004-51324 Supplier Medchemexpress LLC Supplier No. HYW00935410MM/1ML
Di-tert-butyl diisopropylphosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides.
  • It appears as a liquid, colorless to light yellow.
  • It ships at room temperature in the continental US; conditions may vary elsewhere.
  • Store at 4°C under nitrogen.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen.