Learn More
Medchemexpress LLC Di-tert-butyl diisopropylphosphoramidite | 137348-86-8 | 96.9% | 277.39 | 10 G
SDP
Catalog No. 5000451510
Encompass_Preferred
Di-tert-butyl diisopropylphosphoramidite is a phosphorite monomer utilized in the synthesis of oligonucleotides. It is intended for research use only.
  • Used as a phosphorite monomer.
  • Applicable in the synthesis of oligonucleotides.
Catalog No. 50-004-51510 Supplier Medchemexpress LLC Supplier No. HYW00935410G
Di-tert-butyl diisopropylphosphoramidite is a phosphorite monomer utilized in the synthesis of oligonucleotides. It is intended for research use only.
  • Used as a phosphorite monomer.
  • Applicable in the synthesis of oligonucleotides.