Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Di-tert-butyl diisopropylphosphoramidite | 137348-86-8 | MFCD00153506 | 96.9% | 277.39 | 25 G
SDP

Catalog No. 5000366768
Encompass_Preferred

Di-tert-butyl diisopropylphosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides. It is for research use only.

  • Purity of 96.85%
  • Molecular weight of 277.39
  • Chemical formula: C14H32NO2P
  • Appears as a colorless to light yellow liquid
  • Recommended storage at 4°C under nitrogen

Catalog No. 50-003-66768 Supplier Medchemexpress LLC Supplier No. 25GHYW009354
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Di-tert-butyl diisopropylphosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides. It is for research use only.

  • Purity of 96.85%
  • Molecular weight of 277.39
  • Chemical formula: C14H32NO2P
  • Appears as a colorless to light yellow liquid
  • Recommended storage at 4°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.