Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Di-tert-butyl iminodicarboxylate | 51779-32-9 | >=98.0% | C10H19NO4 | 100 G
SDP

Catalog No. 5000433786
Encompass_Preferred

Di-tert-butyl iminodicarboxylate is a chemical compound supplied as a white to off-white solid. It has a molecular formula of C10H19NO4 and a molecular weight of 217.26. The product is for research use only.

  • Appearance: White to off-white solid
  • Purity: ≥98.0%
  • Molecular formula: C10H19NO4
  • Molecular weight: 217.26
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-33786 Supplier Medchemexpress LLC Supplier No. HY60280100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Di-tert-butyl iminodicarboxylate is a chemical compound supplied as a white to off-white solid. It has a molecular formula of C10H19NO4 and a molecular weight of 217.26. The product is for research use only.

  • Appearance: White to off-white solid
  • Purity: ≥98.0%
  • Molecular formula: C10H19NO4
  • Molecular weight: 217.26
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.