Learn More
Medchemexpress LLC Di-tert-butyl iminodicarboxylate | 51779-32-9 | ≥98.0% | C10H19NO4 | 25 G
SDP
Catalog No. 5000358708
Encompass_Preferred
Di-tert-butyl iminodicarboxylate is a biochemical reagent used in life science research. It serves as a biological material or organic compound and is not for patient use. This reagent is key in preparing various compounds, such as diamino-2-butene isomers, di-tert-butyl N-3-butenyliminodicarboxylate, N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate, and tetrafibricin fragments. It also acts as a Modified Gabriel Reagent.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Used in the preparation of diamino-2-butene isomers
  • Used in the preparation of di-tert-butyl N-3-butenyliminodicarboxylate
  • Used in the preparation of N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate
  • Used in the preparation of tetrafibricin fragments (C1-C40)
  • Acts as a Modified Gabriel Reagent
Catalog No. 50-003-58708 Supplier Medchemexpress LLC Supplier No. 25GHY60280
Di-tert-butyl iminodicarboxylate is a biochemical reagent used in life science research. It serves as a biological material or organic compound and is not for patient use. This reagent is key in preparing various compounds, such as diamino-2-butene isomers, di-tert-butyl N-3-butenyliminodicarboxylate, N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate, and tetrafibricin fragments. It also acts as a Modified Gabriel Reagent.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Used in the preparation of diamino-2-butene isomers
  • Used in the preparation of di-tert-butyl N-3-butenyliminodicarboxylate
  • Used in the preparation of N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate
  • Used in the preparation of tetrafibricin fragments (C1-C40)
  • Acts as a Modified Gabriel Reagent