Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-Tert-Butyl N,N-Diisopropylphosphoramidite

Catalog No. 4193625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-936-25G 5 g
41-936-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 41-936-25G Supplier Sigma Aldrich Supplier No. 4193625G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di-tert-butyl N,N-diisopropylphosphoramidite is a general reagent to introduce tert-butyl-protected phosphate groups. It is commonly used in phosphorylation of biomolecules. It can also be used in the synthesis of phosphoramidate-linked glycoconjugates.

Specifications

CAS 137348-86-8
Density 0.879 g/mL at 25°C
Boiling Point 85°C to 90°C (0.2 mmHg, lit.)
Molecular Formula C14H32NO2P
Refractive Index n20/D 1.444
Linear Formula [(CH3)2CH]2NP[OC(CH3)3]2
MDL Number MFCD00153506
Quantity 5 g
Molecular Weight (g/mol) 277.38 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.