Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl phosphite, 96%

Catalog No. AC449390250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449390250 25 g Glass bottle
AC449390010 1 g Glass bottle
AC449390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449390250 Supplier Thermo Scientific Chemicals Supplier No. 449390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13086-84-5 | C8H18O3P | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13086-84-5
Molecular Formula C8H18O3P
Molecular Weight (g/mol) 193.20
MDL Number MFCD00014999,MFCD00014999
InChI Key GEBLOQXLELCEEO-UHFFFAOYSA-N
Synonym di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester
PubChem CID 21959072
IUPAC Name ditert-butyl phosphite
SMILES CC(C)(C)O[P+](=O)OC(C)(C)C

Specifications

Density 0.9950g/mL
Boiling Point 66°C to 68°C (0.5 mmHg)
Flash Point 163°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3CO]2P(O)H
Quantity 25 g
Specific Gravity 0.995
Solubility Information Solubility in water: immiscible
Formula Weight 193.2
Percent Purity 96%
Chemical Name or Material Di-tert-butyl phosphite
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
EINECSNumber 235-996-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.