Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-Tert-Butylchlorophosphine

Catalog No. 30155825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-155-825G 25 g
30-155-85G 5 g
30-155-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 30-155-825G Supplier Sigma Aldrich Supplier No. 30155825G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bulky ligand used with Pd(OAc)2 in efficient Suzuki-Miyaura cross-couplings of arylboronic acids with aryl bromides and chlorides.

Specifications

CAS 13716-10-4
Density 0.951 g/mL at 25°C
Boiling Point 48°C (3 mmHg, lit.)
Molecular Formula C8H18ClP
Refractive Index n20/D 1.482
Linear Formula [(CH3)3C]2PCl
MDL Number MFCD00008815
Quantity 25 g
Molecular Weight (g/mol) 180.66 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.