Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butylchlorophosphine, 96%

Catalog No. AAB2099814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2099814 25 g
AAB2099806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2099814 Supplier Thermo Scientific Chemicals Supplier No. B2099814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13716-10-4 | C8H18ClP | 180.656 g/mol

Di-tert-butylchlorophosphine is associated with palladium(II) acetate and used in Suzuki-Miyaura cross-couplings of arylboronic acids with aryl bromides and chlorides. It serves as a ligand and used in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations. Further, it is used in asymmetric and transition metal coupling reactions. In addition to this, it plays an important role in the preparation of other phosphines in catalytic polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butylchlorophosphine is associated with palladium(II) acetate and used in Suzuki-Miyaura cross-couplings of arylboronic acids with aryl bromides and chlorides. It serves as a ligand and used in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations. Further, it is used in asymmetric and transition metal coupling reactions. In addition to this, it plays an important role in the preparation of other phosphines in catalytic polymerization reactions.

Solubility
Miscible with terahydrofuran.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13716-10-4
Molecular Formula C8H18ClP
Molecular Weight (g/mol) 180.656
MDL Number MFCD00008815
InChI Key MCRSZLVSRGTMIH-UHFFFAOYSA-N
Synonym di-tert-butylchlorophosphine, di-tert-butylchlorophosphane, di-t-butylchlorophosphine, phosphinous chloride, bis 1,1-dimethylethyl, chlorodi-tert-butylphosphine, phosphinous chloride, di-tert-butyl, di-tert-butyl chloro phosphane, di-tert-butylphosphinous chloride, clptbu2, pubchem6477
PubChem CID 139566
IUPAC Name ditert-butyl(chloro)phosphane
SMILES CC(C)(C)P(C(C)(C)C)Cl

Specifications

Density 0.951
Boiling Point 47°C to 49°C (3 mmHg)
Flash Point 62°C (144°F)
Odor Acrid
Refractive Index 1.484
Linear Formula [(CH3)3C]2PCl
Quantity 25 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Miscible with terahydrofuran.
Formula Weight 180.66
Percent Purity 96%
Chemical Name or Material Di-tert-butylchlorophosphine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.