Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-Butylsilane 95.0+%, TCI America™
SDP

Catalog No. D37315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37315G 5 g
D37311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37315G Supplier TCI America Supplier No. D37315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30736-07-3
Molecular Formula C8H20Si
MDL Number MFCD00270996

Specifications

Melting Point -38°C
Color Yellow
Boiling Point 129°C
Quantity 5 g
UN Number 1993
Formula Weight 144.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-Butylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.