Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-Tert-Butylsilyl Bis(Trifluoromethanesulfonate)

Catalog No. 26202125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-202-125G 25 g
21-500-725ML 5 g
26-202-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 26-202-125G Supplier Sigma Aldrich Supplier No. 26202125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protecting group reagent for 1,3-diols used recently in a synthesis of N-homoceramides. Also used for selective +a-galactosylation in the synthesis of +a-galactosyl ceramides.

Specifications

CAS 85272-31-7
Density 1.352 g/mL at 25°C
Boiling Point 73°C to 75°C (0.35 mmHg, lit.)
Molecular Formula C10H18F6O6S2Si
Linear Formula (CF3SO3)2Si[C(CH3)3]2
Refractive Index n20/D 1.398
MDL Number MFCD00010581
Quantity 25 g
Synonym DTBS ditriflate; Di-tert-butylsilyl ditriflate; Trifluoromethanesulfonic acid di-tert-butylsilylene ester
Molecular Weight (g/mol) 440.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.