Learn More
Andwin Scientific DI THERMAL CRYO 38X13 WH 750PK
SDP
Catalog No. 50727696
Encompass_Preferred
The Direct Thermal CryoTags feature compatiblity with a wide range of temperatures from -196C to 70C and compatiblity with tubes labeled with Direct Thermal CryoTags. This product is economical as it reduces label waste by printing only the quantity desired. The labels can be printed from direct thermal printers such as Dymo LabelWriters.. This product measures 1.50" x 0.50" (38x13mm), which is ideal for use with 1.5 mL tubes. It is sold in a pack of 750.
Catalog No. 50-727-696 Supplier Andwin Scientific Supplier No. 775772
The Direct Thermal CryoTags feature compatiblity with a wide range of temperatures from -196C to 70C and compatiblity with tubes labeled with Direct Thermal CryoTags. This product is economical as it reduces label waste by printing only the quantity desired. The labels can be printed from direct thermal printers such as Dymo LabelWriters.. This product measures 1.50" x 0.50" (38x13mm), which is ideal for use with 1.5 mL tubes. It is sold in a pack of 750.