Learn More
Andwin Scientific DI THERMAL CRYO 51 x 6 White, 1000pack
SDP
Catalog No. 50727701
Encompass_Preferred
Compatible with direct thermal printers, the 51Z6mm Cryo-Tags in White are used on microplates. They are temperature resistant from -196-70 deg.C and freezable in liquid nitrogen. It comes in 1000 a pack.
Catalog No. 50-727-701 Supplier Andwin Scientific Supplier No. 775775
Compatible with direct thermal printers, the 51Z6mm Cryo-Tags in White are used on microplates. They are temperature resistant from -196-70 deg.C and freezable in liquid nitrogen. It comes in 1000 a pack.