Learn More
Enzo Life Sciences Di-ubiquitin (K63-linked) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW07300050
Encompass_Preferred
Polyubiquitin chains, linked through specific lysine residues, are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins. Polyubiquitin chains, linked through specific lysine residues, are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins. Alternative name: Ub2. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM TRIS-HCl, pH 7.5, containing 0.15M NaCl, 1mM EDTA. Source: Protein components produced in E. coli. Synthesised enzymatically in vitro. Long Term Storage: -20°C.
Catalog No. 50-201-2264