Learn More
Enzo Life Sciences Di-ubiquitin (linear) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW07750100
Encompass_Preferred
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with ubiquitin binding domain (UBDs) containing proteins. Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with ubiquitin binding domain (UBDs) containing proteins. Alternative name: Ub2. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, pH 8.0, containing 50mM sodium chloride. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-2265