Learn More
Medchemexpress LLC Di((Z)-non-2-en-1-yl) 8,8'-((((2-(dimethylamino)ethyl)thio)carbonyl)azanediyl)dioctanoate | 1777792-34-3 | 98.0% | C39H72N2O5S | 10MG
SDP
Catalog No. 5000261467
Encompass_Preferred
ATX-002 is an ionizable cationic lipid intended for incorporation into lipid nanoparticles to enable delivery of RNA therapeutics. It is described in patent literature and provided by research suppliers with reported properties useful for formulation and preclinical studies.
  • Designed for lipid nanoparticle encapsulation of RNA.
  • Calculated pKa 8.68 and measured pKa 6.03, enabling pH-dependent ionization.
  • Oil-phase material, light yellow to yellow in appearance.
  • High purity suitable for preclinical research (reported 98.0%).
  • Molecular formula C39H72N2O5S; exact mass ≈ 681.06.
  • Used in siRNA and mRNA formulation studies with demonstrated in vivo activity in patent examples.
Catalog No. 50-002-61467 Supplier Medchemexpress LLC Supplier No. HY14265410MG
ATX-002 is an ionizable cationic lipid intended for incorporation into lipid nanoparticles to enable delivery of RNA therapeutics. It is described in patent literature and provided by research suppliers with reported properties useful for formulation and preclinical studies.
  • Designed for lipid nanoparticle encapsulation of RNA.
  • Calculated pKa 8.68 and measured pKa 6.03, enabling pH-dependent ionization.
  • Oil-phase material, light yellow to yellow in appearance.
  • High purity suitable for preclinical research (reported 98.0%).
  • Molecular formula C39H72N2O5S; exact mass ≈ 681.06.
  • Used in siRNA and mRNA formulation studies with demonstrated in vivo activity in patent examples.