Learn More
Medchemexpress LLC Di((Z)-non-2-en-1-yl) 9-((4-(dimethylamino)butanoyl)oxy)heptadecanedioate | 1351586-50-9 | 98.0% | 678.04 g·mol⁻¹ | C41H75NO6 | 5 MG
SDP
Catalog No. 5000221446
Encompass_Preferred
L319 is an ionizable cationic lipidoid (Di((Z)-non-2-en-1-yl) 9-((4-(dimethylamino)butanoyl)oxy)heptadecanedioate) used to formulate synthetic liposomes and lipid nanoparticles. The material is provided at research-grade purity and is intended for formulation and delivery studies in preclinical research.
  • Ionizable cationic structure suitable for pH-dependent delivery.
  • High supplied purity (98.0%) for reproducible formulation.
  • Applicable to synthetic liposome and lipid nanoparticle assembly.
  • Well-characterized molecular properties facilitate formulation optimization.
  • Available in small research quantities for screening and development.
Catalog No. 50-002-21446 Supplier Medchemexpress LLC Supplier No. HY1392985MG
L319 is an ionizable cationic lipidoid (Di((Z)-non-2-en-1-yl) 9-((4-(dimethylamino)butanoyl)oxy)heptadecanedioate) used to formulate synthetic liposomes and lipid nanoparticles. The material is provided at research-grade purity and is intended for formulation and delivery studies in preclinical research.
  • Ionizable cationic structure suitable for pH-dependent delivery.
  • High supplied purity (98.0%) for reproducible formulation.
  • Applicable to synthetic liposome and lipid nanoparticle assembly.
  • Well-characterized molecular properties facilitate formulation optimization.
  • Available in small research quantities for screening and development.