Learn More
Medchemexpress LLC Di((Z)-non-2-en-1-yl) 9-((4-(dimethylamino)butanoyl)oxy)heptadecanedioate | 1351586-50-9 | 98.0% | C41H75NO6 | 10MG

Supplier: Medchemexpress LLC HY13929810MG
L319 is an ionizable cationic lipidoid used to formulate synthetic liposomes and lipid nanoparticles for in vitro research. Described in patent WO-2011153493-A2 (compound 1), it is provided as a high-purity research chemical.
- Ionizable cationic lipidoid suitable for liposome and lipid nanoparticle formulation.
- High purity (98.0%) for research applications.
- Molecular weight: 678.04 g/mol; formula: C41H75NO6.
- Appearance: colorless to light yellow liquid.
- Storage stability: stable at -20°C for extended storage in pure form.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.