Learn More
Biotium DIA
Catalog No. 501964718
Encompass_Preferred
DiA 4-(4-Dihexadecylaminostyryl)-N-methylpyridinium iodide) is a green fluorescent membrane dye which diffuses much faster than DiO in cell membranes. DiA and DiI have been used together for two color membrane staining. Properties: Red solid soluble in DMSO; Store at 4°C and protect from light, especially in solution; C46H79IN2; MW: 787; [114041-00-8]
Catalog No. 50-196-4718 Supplier Biotium Supplier No. 60013
DiA 4-(4-Dihexadecylaminostyryl)-N-methylpyridinium iodide) is a green fluorescent membrane dye which diffuses much faster than DiO in cell membranes. DiA and DiI have been used together for two color membrane staining. Properties: Red solid soluble in DMSO; Store at 4°C and protect from light, especially in solution; C46H79IN2; MW: 787; [114041-00-8]