Learn More
Grainger General Purpose Diagonal Cutting Pliers, Flush Cut Profile
Catalog No. NC2314556
Encompass_Preferred
The Grainger 6" Flush Cut Diagonal Cutting Pliers are general purpose pliers designed for cutting wire and small bolts. They feature induction hardened cutting edges for durability and have a flush cutting profile that provides a smooth, even cut. The pliers have serrated jaws for gripping and are made from forged chrome vanadium steel. They have plastic dipped handles for comfort and come individually packaged. These are quality pliers suitable for general purpose cutting applications.
Catalog No. NC2314556 Supplier Grainger Supplier No. 5EPR6
The Grainger 6" Flush Cut Diagonal Cutting Pliers are general purpose pliers designed for cutting wire and small bolts. They feature induction hardened cutting edges for durability and have a flush cutting profile that provides a smooth, even cut. The pliers have serrated jaws for gripping and are made from forged chrome vanadium steel. They have plastic dipped handles for comfort and come individually packaged. These are quality pliers suitable for general purpose cutting applications.