Learn More
Mercodia Inc Diabetes Antigen Control, Human, Low High, Unassayed, For Insulin C-Peptide Iso-Insulin Proinsulin
SDP
Catalog No. NC9597840
Encompass_Preferred
Mercodia Diabetes Antigen Control is designed to be used as a two-level control for human insulin, C-peptide and proinsulin. It is manufactured from human serum and human insulin, C-peptide and proinsulin.
Catalog No. NC9597840 Supplier Mercodia Inc Supplier No. 10113401
Mercodia Diabetes Antigen Control is designed to be used as a two-level control for human insulin, C-peptide and proinsulin. It is manufactured from human serum and human insulin, C-peptide and proinsulin.