Learn More
Mercodia Inc Diabetes Sample Buffer, For Insulin and Proinsulin ELISAs
SDP
Catalog No. NC9882274
Encompass_Preferred
Mercodia Diabetes Sample Buffer is a dilution buffer that can be used to dilute samples in Mercodia ELISA kits within our diabetes assay portfolio. This includes human, rodent, and animal insulin assays. This dilution buffer is comparable to the 0 or blank calibrator found in each assay kit.
Catalog No. NC9882274 Supplier Mercodia Inc Supplier No. 10119501
Mercodia Diabetes Sample Buffer is a dilution buffer that can be used to dilute samples in Mercodia ELISA kits within our diabetes assay portfolio. This includes human, rodent, and animal insulin assays. This dilution buffer is comparable to the 0 or blank calibrator found in each assay kit.