Learn More
Invivogen diABZI (Compound 3): Potent Non-CDN STING Agonist, Increased Bioavailability, Validated Purity, 2 mg
SDP
Catalog No. NC2683249
Encompass
diABZI is a potent non-canonical STING agonist that activates NF-κB and IRF pathways, enhancing immune responses. It offers increased bioavailability compared to classical STING agonists, making it a potential therapeutic for cancer and viral infections, such as SARS-CoV-2. With validated purity and precise working concentration (0.01 - 30 µM), diABZI can effectively induce type-I interferons and pro-inflammatory cytokines, providing promising treatment avenues for various conditions.
Catalog No. NC2683249 Supplier Invivogen Supplier No. TLRLDIABZI2
diABZI is a potent non-canonical STING agonist that activates NF-κB and IRF pathways, enhancing immune responses. It offers increased bioavailability compared to classical STING agonists, making it a potential therapeutic for cancer and viral infections, such as SARS-CoV-2. With validated purity and precise working concentration (0.01 - 30 µM), diABZI can effectively induce type-I interferons and pro-inflammatory cytokines, providing promising treatment avenues for various conditions.