Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DiABZI-C2-NH2 | 2137975-93-8 | 96.1% | C36H43N13O4 | 10MG
SDP

Catalog No. 5000261440
Encompass_Preferred

diABZI-C2-NH2 is a synthetic small-molecule STING agonist for research use. It is an active analogue containing a primary amine functionality and is supplied with supporting documentation (data sheet, COA, SDS) for experimental reproducibility.

  • Acts as a STING agonist in immuno-oncology research
  • Contains a primary amine functionality
  • High purity: 96.1%
  • Available as solid and as a 10 mM solution in DMSO
  • Soluble in DMSO (100 mg/mL) for in vitro use
  • Recommended storage: solid at -20°C under nitrogen; in solution store at -80°C

Catalog No. 50-002-61440 Supplier Medchemexpress LLC Supplier No. HY13732010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

diABZI-C2-NH2 is a synthetic small-molecule STING agonist for research use. It is an active analogue containing a primary amine functionality and is supplied with supporting documentation (data sheet, COA, SDS) for experimental reproducibility.

  • Acts as a STING agonist in immuno-oncology research
  • Contains a primary amine functionality
  • High purity: 96.1%
  • Available as solid and as a 10 mM solution in DMSO
  • Soluble in DMSO (100 mg/mL) for in vitro use
  • Recommended storage: solid at -20°C under nitrogen; in solution store at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.