Learn More
Medchemexpress LLC DiABZI-C2-NH2 | 2137975-93-8 | 96.14% | 721.81 | 100 MG
SDP
Catalog No. 5000348622
Encompass_Preferred
diABZI-C2-NH2 is an active analogue containing a primary amine functionality and acts as a STING agonist. It has shown strong anti-tumor activity in immunocompetent mice with syngeneic colon tumors, leading to complete and lasting regression of tumors. This compound can be covalently immobilized on sepharose beads for affinity-capture of potential target proteins from THP1 cell lysate.
  • Functions as a STING agonist
  • Exhibits strong anti-tumor activity in immunocompetent mice
  • Leads to complete and lasting regression of tumors
  • Can be covalently immobilized on sepharose beads
  • Useful for affinity-capture of potential target proteins from THP1 cell lysate
  • Enhanced binding to STING and cellular function
Catalog No. 50-003-48622 Supplier Medchemexpress LLC Supplier No. HY137320100MG
diABZI-C2-NH2 is an active analogue containing a primary amine functionality and acts as a STING agonist. It has shown strong anti-tumor activity in immunocompetent mice with syngeneic colon tumors, leading to complete and lasting regression of tumors. This compound can be covalently immobilized on sepharose beads for affinity-capture of potential target proteins from THP1 cell lysate.
  • Functions as a STING agonist
  • Exhibits strong anti-tumor activity in immunocompetent mice
  • Leads to complete and lasting regression of tumors
  • Can be covalently immobilized on sepharose beads
  • Useful for affinity-capture of potential target proteins from THP1 cell lysate
  • Enhanced binding to STING and cellular function