Learn More
Medchemexpress LLC HY-137320 5mg , diABZI-C2-NH2 CAS:2137975-93-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1373205MG
Encompass_Preferred
HY-137320 5mg diABZI-C2-NH2 CAS: 2137975-93-8 diABZI-C2-NH2, an active analogue containing a primary amine functionality, is a STING agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1800