Learn More
Medchemexpress LLC Diabzi sting agonist-1 | 2138498-18-5 | MFCD31746907 | 99.9% | 849.94 g/mol | C42H51N13O7 | 5 MG
SDP
Catalog No. 5000262417
Encompass_Preferred
diABZI STING agonist-1 (tautomerism) is a non-nucleotide small-molecule agonist of the STING (stimulator of interferon genes) signaling pathway. It activates STING with reported EC50 values of 130 nM (human) and 186 nM (mouse), is supplied for research use only, and is provided in a small-scale research vial.
  • Potent STING receptor agonist with low-nanomolar activity
  • Reported EC50 values: human 130 nM, mouse 186 nM
  • High chemical purity (99.9%)
  • Molecular weight 849.94 g/mol; formula C42H51N13O7
  • Supplied as a 5 mg research vial
  • Used in studies demonstrating STING-dependent activation in vivo and in vitro
Catalog No. 50-002-62417 Supplier Medchemexpress LLC Supplier No. HY1129215MG
diABZI STING agonist-1 (tautomerism) is a non-nucleotide small-molecule agonist of the STING (stimulator of interferon genes) signaling pathway. It activates STING with reported EC50 values of 130 nM (human) and 186 nM (mouse), is supplied for research use only, and is provided in a small-scale research vial.
  • Potent STING receptor agonist with low-nanomolar activity
  • Reported EC50 values: human 130 nM, mouse 186 nM
  • High chemical purity (99.9%)
  • Molecular weight 849.94 g/mol; formula C42H51N13O7
  • Supplied as a 5 mg research vial
  • Used in studies demonstrating STING-dependent activation in vivo and in vitro