Learn More
Medchemexpress LLC HY-112921A 5mg , diABZI STING agonist-1 CAS:2138299-33-7 Purity:>98%
SDP
Catalog No. 502261667
Encompass_Preferred
HY-112921A 5mg diABZI STING agonist-1 CAS: 2138299-33-7 diABZI STING agonist-1 is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130, 186 nM for human and mouse, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1667 Supplier Medchemexpress LLC Supplier No. HY112921A5MG
HY-112921A 5mg diABZI STING agonist-1 CAS: 2138299-33-7 diABZI STING agonist-1 is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130, 186 nM for human and mouse, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.