Learn More
TARGETMOL CHEMICALS INC DIABZI STING AGONIST-1 T 5MG
SDP
Catalog No. 502581587
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. diABZI STING agonist-1 (Tautomerism) (diABZI STING agonist (Compound 3)) is a selective stimulator of interferon genes (STING) receptor agonist with EC50s of 130 186 nM for human and mouse respectively. purity: 99%
Catalog No. 50-258-1587 Supplier TARGETMOL CHEMICALS INC Supplier No. T110355MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. diABZI STING agonist-1 (Tautomerism) (diABZI STING agonist (Compound 3)) is a selective stimulator of interferon genes (STING) receptor agonist with EC50s of 130 186 nM for human and mouse respectively. purity: 99%