Learn More
Medchemexpress LLC DIABZI STING AGONIST-1 TRIHYDR
SDP
Catalog No. NC1743833
Encompass_Preferred
diABZI STING agonist-1 (trihydrochloride) is a selective stimulator of interferon genes (STING) receptor agonist, with an EC50s of 130 for human PBMCs. CAS # 2138299-34-8
Catalog No. NC1743833 Supplier Medchemexpress LLC Supplier No. HY112921B10MG
diABZI STING agonist-1 (trihydrochloride) is a selective stimulator of interferon genes (STING) receptor agonist, with an EC50s of 130 for human PBMCs. CAS # 2138299-34-8