Learn More
Medchemexpress LLC HY-N0283 50mg Medchemexpress, Diacerein CAS:13739-02-1 Purity:>98%
SDP
Catalog No. 501933506
Encompass_Preferred
Medchemexpress, HY-N0283 50mg Diacerein CAS:13739-02-1 Diacerein (Diacerhein), a interleukin-1 beta inhibitor, is a slow-acting medicine of the class anthraquinone used to treat joint diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3506 Supplier Medchemexpress LLC Supplier No. HYN028350MG
Medchemexpress, HY-N0283 50mg Diacerein CAS:13739-02-1 Diacerein (Diacerhein), a interleukin-1 beta inhibitor, is a slow-acting medicine of the class anthraquinone used to treat joint diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.