Learn More
Apexbio Technology LLC Diacerein 13739-02-1 50mg
SDP
Catalog No. 502240456
Encompass_Preferred
Diacerein (CAS 13739-02-1) is an orally available anthraquinone derivative that modulates inflammatory signaling by interfering with interleukin-1 (IL-1) pathways It acts primarily by inhibiting the synthesis of IL-1 converting enzyme thereby reducing the activation of IL-1 and subsequently attenuating downstream inflammatory cascades Diacerein has been employed in research investigating the mechanisms of inflammation and cartilage degradation associated with osteoarthritis Additionally in animal models it has demonstrated the ability to mitigate airway hyperresponsiveness and local inflammation supporting its use in exploring the pathophysiology of bronchial asthma
This item is not returnable. View return policy
Catalog No. 50-224-0456 Supplier Apexbio Technology LLC Supplier No. B172650
Diacerein (CAS 13739-02-1) is an orally available anthraquinone derivative that modulates inflammatory signaling by interfering with interleukin-1 (IL-1) pathways It acts primarily by inhibiting the synthesis of IL-1 converting enzyme thereby reducing the activation of IL-1 and subsequently attenuating downstream inflammatory cascades Diacerein has been employed in research investigating the mechanisms of inflammation and cartilage degradation associated with osteoarthritis Additionally in animal models it has demonstrated the ability to mitigate airway hyperresponsiveness and local inflammation supporting its use in exploring the pathophysiology of bronchial asthma