Learn More
Strem, An Ascensus Company CAS# 373650-12-5. 250mg. Diacetato[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II) Ru(OAc)2[(S)-segphos]. MFCD09753024
SDP
Supplier:  Strem, An Ascensus Company 440169250MG
Encompass
CAS# 373650-12-5. 250mg. Diacetato[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II) Ru(OAc)2[(S)-segphos]. MFCD09753024. Molecular Weight: 829.73. Molecular Formula: C42H34O8P2Ru. Color/form: yellow to black solid Strem# 44-0169
Catalog No. 50-901-11841