Learn More
Medchemexpress LLC HY-B2187 100mg Medchemexpress, Diacetazotol CAS:83-63-6 Purity:>98%
SDP
Catalog No. 501933100
Encompass_Preferred
Medchemexpress, HY-B2187 100mg Diacetazotol CAS:83-63-6 Diacetazotol inhibits dioxin-induced ethoxyresorufin-O-deethylase ( EROD ) activity with IC 50 of 75±4 nM. Diacetazotol extracts from patent US20070032458, compound 3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3100 Supplier Medchemexpress LLC Supplier No. HYB2187100MG
Medchemexpress, HY-B2187 100mg Diacetazotol CAS:83-63-6 Diacetazotol inhibits dioxin-induced ethoxyresorufin-O-deethylase ( EROD ) activity with IC 50 of 75±4 nM. Diacetazotol extracts from patent US20070032458, compound 3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.