Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetin (contains Mono-, Tri-, Glycerol), TCI America™
SDP

Catalog No. G007725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G007725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G007725G Supplier TCI America Supplier No. G007725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25395-31-7
Molecular Formula C7H12O5
MDL Number MFCD00008717
Synonym glyceryl diacetate, 1,2-diacetin, glycerol diacetate, 1,2,3-propanetriol, diacetate, 3-hydroxypropane-1,2-diyl diacetate, glycerol 1,2-diacetate, 1,2-diacylglycerol, 2,3-diacetin, 1,2-diacetylglycerol, acetin, di

Specifications

Color Yellow
Boiling Point 261°C
Quantity 25 g
Formula Weight 176.17
Physical Form Liquid
Chemical Name or Material Diacetin (contains Mono-, Tri-, Glycerol)
EINECSNumber (2)-0666&(2)-0753
RTECSNumber AK3325000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.