Learn More
Cayman Chemical diacetoxy Scirpenol, Purity: ≥98%, CAS: 2270-40-8, 10 mg
SDP
Catalog No. 502951473
Encompass_Preferred
A trichothecene mycotoxin; is toxic to pigs (LD50 = 0.38 mg/kg, i.v.); is lethal to brine shrimp (LC50 = 0.39 µg/ml); suppresses phagocytosis by, and superoxide production in, isolated microphages in an S. cerevisiae model of phagocytosis at 1 µg/ml; is toxic to barley, wheat, and sorghum seedlings (LD50s = 1.26, 3.98, and 10 mg/L, respectively); has been found as a contaminant in grasses intended to be used as animal feed
Catalog No. 50-295-1473 Supplier Cayman Chemical Supplier No. 1142710 MG
A trichothecene mycotoxin; is toxic to pigs (LD50 = 0.38 mg/kg, i.v.); is lethal to brine shrimp (LC50 = 0.39 µg/ml); suppresses phagocytosis by, and superoxide production in, isolated microphages in an S. cerevisiae model of phagocytosis at 1 µg/ml; is toxic to barley, wheat, and sorghum seedlings (LD50s = 1.26, 3.98, and 10 mg/L, respectively); has been found as a contaminant in grasses intended to be used as animal feed