Learn More
MSC Xcelite Diagonal Cutting Plier, 0.8 mm/20 AWG Capacity, 1 ea
Catalog No. 501920551
Encompass
This diagonal cutting plier is designed for precise cutting tasks, offering a 0.8 mm and 20 AWG cutting capacity. It features a diagonal head and a 15/32 inch jaw length, suitable for various applications.
  • Cutting capacity: 0.8 mm and 20 AWG
  • Diagonal head style
  • Steel joint ensures durability
  • Perfect cutting edge for precise results
  • Durable construction
  • Oval head design
  • Precise tip cutting capability
  • High quality
  • Cushion grip handle
  • ESD safe
Catalog No. 50-192-0551 Supplier MSC Supplier No. 38632600
This diagonal cutting plier is designed for precise cutting tasks, offering a 0.8 mm and 20 AWG cutting capacity. It features a diagonal head and a 15/32 inch jaw length, suitable for various applications.
  • Cutting capacity: 0.8 mm and 20 AWG
  • Diagonal head style
  • Steel joint ensures durability
  • Perfect cutting edge for precise results
  • Durable construction
  • Oval head design
  • Precise tip cutting capability
  • High quality
  • Cushion grip handle
  • ESD safe