Learn More
Diagenode Inc dPLEX Small RNA-Seq Kit, for Illumina sequencing, 24 reactions, ligation-free, 10 pg to 10 ng small RNA input, 100 pg to 100 ng total RNA input, single tube protocol, 5 hours preparation time
Catalog No. NC2469623
Encompass
dPLEX Small RNA-Seq Kit, for Illumina sequencing, 24 reactions, ligation-free, 10 pg to 10 ng small RNA input, 100 pg to 100 ng total RNA input, single tube protocol, 5 hours preparation time
Catalog No. NC2469623 Supplier Diagenode Inc Supplier No. C05030001
dPLEX Small RNA-Seq Kit, for Illumina sequencing, 24 reactions, ligation-free, 10 pg to 10 ng small RNA input, 100 pg to 100 ng total RNA input, single tube protocol, 5 hours preparation time