Learn More
Diagenode Inc DIAGMAG02 MAGNETIC RACK 16X0.2
Catalog No. NC0207210
Encompass
The DiaMag02 is a powerful magnet which has been designed for controlled and rapid isolation of your DNA bound to magnetic beads. It allows for processing 16 samples at a time.
Catalog No. NC0207210 Supplier Diagenode Inc Supplier No. B04000001
The DiaMag02 is a powerful magnet which has been designed for controlled and rapid isolation of your DNA bound to magnetic beads. It allows for processing 16 samples at a time.